Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FSCN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121975
This item is not returnable.
View return policy
Description
FSCN2 acts as an actin bundling protein. May play a pivotal role in photoreceptor cell-specific events, such as disk morphogenesis.
Specifications
FSCN2 | |
Polyclonal | |
Unconjugated | |
FSCN2 | |
A930022G03; Ahl8; C630046B20Rik; fascin 2; fascin 2, retinal; fascin actin-bundling protein 2, retinal; fascin homolog 2, actin-bundling protein, retinal; fascin homolog 2, actin-bundling protein, retinal (Strongylocentrotus purpuratus); fascin-2; Fscn2; Retinal fascin; RFSN; RP30 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
25794 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
O14926 | |
FSCN2 | |
Gln8-His258 (Accession O14926), with N-terminal His Tag. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction