Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FSCN3/Fascin-3 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11285R

Description
FSCN3 (Fascin Actin-Bundling Protein 3) is a Protein Coding gene. Among its related pathways are MAPK-Erk Pathway. Gene Ontology (GO) annotations related to this gene include actin filament binding and protein binding, bridging. An important paralog of this gene is FSCN1.Specifications
FSCN3/Fascin-3 | |
Polyclonal | |
Unconjugated | |
FSCN3 | |
fascin (testis); fascin 3; fascin actin-bundling protein 3; fascin actin-bundling protein 3, testicular; fascin homolog 3, actin-bundling protein, testicular; fascin homolog 3, actin-bundling protein, testicular (Strongylocentrotus purpuratus); Fascin-3; FSCN3; testicular secretory protein Li 18; Testis fascin | |
Rabbit | |
Protein A | |
RUO | |
29999 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9NQT6 | |
FSCN3 | |
KLH conjugated synthetic peptide derived from human Fascin-3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction