Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FSD1 Monoclonal Antibody (1G3)
GREENER_CHOICE

Catalog No. PIMA522569
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522569 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522569 Supplier Invitrogen™ Supplier No. MA522569
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MPDEDSKIDH YVLEYRRTNF EGPPRLKEDQ PWMVIEGIRQ TEYTLTGLKF DMKYMNFRVK ACNKAVAGEF SEPVTLETPA FMFRLDASTS HQNLRVDDLS VEWDAMGGKV QDIKAREKDG KGRTASPINS PARGTPSPKR MPSGRGGRDR FTAESYTVLG DTLIDGGEHY WEVRYEPDSK AFGVGVAYRS LGRFEQLGKT AASWCLHVNN WLQVSFTAKH ANKVKVLDAP VPDCLGVHCD FHQGLLSFYN ARTKQ.

FSD1 encodes a centrosome associated protein that is characterized by an N-terminal coiled-coil region downstream of B-box (BBC) domain, a central fibronectin type III domain, and a C-terminal repeats in splA and RyR (SPRY) domain. The encoded protein associates with a subset of microtubules and may be involved in the stability and organization of microtubules during cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSD1
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone 1G3
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene FSD1
Gene Accession No. Q9BTV5
Gene Alias fibronectin type 3 and SPRY (spla, ryanodine) domain containing (with coiled-coil motif) 1; fibronectin type 3 and SPRY domain containing 1; fibronectin type 3 and SPRY domain-containing protein; fibronectin type III and SPRY domain containing 1; fibronectin type III and SPRY domain-containing protein 1; Fsd1; GLFND; Microtubule-associated protein GLFND; MID1-related protein 1; midline 1-related protein 1; MIR1; VLP27
Gene Symbols FSD1
Host Species Mouse
Immunogen FSD1 (-, 1 a.a. ~ 309 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79187
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.