Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FSD1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142208
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
FSD1 encodes a centrosome associated protein that is characterized by an N-terminal coiled-coil region downstream of B-box (BBC) domain, a central fibronectin type III domain, and a C-terminal repeats in splA and RyR (SPRY) domain. The encoded protein associates with a subset of microtubules and may be involved in the stability and organization of microtubules during cytokinesis.
Specifications
| FSD1 | |
| Polyclonal | |
| Unconjugated | |
| FSD1 | |
| fibronectin type 3 and SPRY (spla, ryanodine) domain containing (with coiled-coil motif) 1; fibronectin type 3 and SPRY domain containing 1; fibronectin type 3 and SPRY domain-containing protein; fibronectin type III and SPRY domain containing 1; fibronectin type III and SPRY domain-containing protein 1; Fsd1; GLFND; Microtubule-associated protein GLFND; MID1-related protein 1; midline 1-related protein 1; MIR1; VLP27 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 79187 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BTV5 | |
| FSD1 | |
| Peptide with sequence C-KVQDIKAREKDGKGR. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction