Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FSD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560255 Supplier Invitrogen™ Supplier No. PA560255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVAGEFSEPV TLETPAFMFR LDASTSHQNL RVDDLSVEWD AMGGKVQDIK AREKDGKGRT ASPINSPARG TPSPKRMPSG RGGRDRFTAE Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

FSD1 encodes a centrosome associated protein that is characterized by an N-terminal coiled-coil region downstream of B-box (BBC) domain, a central fibronectin type III domain, and a C-terminal repeats in splA and RyR (SPRY) domain. The encoded protein associates with a subset of microtubules and may be involved in the stability and organization of microtubules during cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FSD1
Gene Accession No. Q9BTV5
Gene Alias fibronectin type 3 and SPRY (spla, ryanodine) domain containing (with coiled-coil motif) 1; fibronectin type 3 and SPRY domain containing 1; fibronectin type 3 and SPRY domain-containing protein; fibronectin type III and SPRY domain containing 1; fibronectin type III and SPRY domain-containing protein 1; Fsd1; GLFND; Microtubule-associated protein GLFND; MID1-related protein 1; midline 1-related protein 1; MIR1; VLP27
Gene Symbols FSD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FSD1. Recombinant protein control fragment (Product #RP-98974).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79187
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.