Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FSD1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570490
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.
FSD1L is a protein coding gene.
Specifications
FSD1L | |
Polyclonal | |
Unconjugated | |
FSD1L | |
A230072O16Rik; Ccdc10; coiled-coil domain containing 10; coiled-coil domain-containing protein 10; CSDUFD1; cystatin and DUF19 domain containing 1; fibronectin type III and SPRY domain containing 1 like; fibronectin type III and SPRY domain containing 1-like; FSD1 C-terminal like; FSD1 N-terminal like; FSD1 N-terminal-like protein; FSD1CL; FSD1L; FSD1-like protein; Fsd1nl; MIR1; OTTMUSG00000007002; RGD1564809; sb:eu990 | |
Rabbit | |
Affinity chromatography | |
RUO | |
83856 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BXM9 | |
FSD1L | |
synthetic peptide directed towards the C terminal region of human FSD1L. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction