Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSD2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982715
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2715 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2715 Supplier Bioss Supplier No. BS13222R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein that belongs to the FN3/SPRY family of proteins. Alternate splicing results in multiple transcript variants.

Specifications

Antigen FSD2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FSD2
Gene Accession No. A1L4K1
Gene Alias 9830160G03Rik; fibronectin type III and SPRY domain containing 2; Fibronectin type III and SPRY domain-containing protein 2; Fsd2; Minispryn; RGD1305167; RP11-127F21; SPRY domain containing 1; SPRY domain-containing protein 1; Spryd1
Gene Symbols FSD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FSD2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123722
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.