Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSH b-subunit Monoclonal Antibody (ABM601), Abbomax

Catalog No. 502204151
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4151 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4151 Supplier Abbomax Inc Supplier No. 603170
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Human pituitary Cellular location: Secreted.

FSH b-subunit Monoclonal antibody specifically detects FSH b-subunit in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen FSH b-subunit
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ABM601
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene FSHB
Gene Accession No. P01225
Gene Alias follicle stimulating hormone beta subunit; follicle stimulating hormone, beta polypeptide; follicle-stimulating hormone beta subunit; Follitropin beta chain; follitropin subunit beta; follitropin, beta chain; FSHB; FSH-B; FSH-beta; HH24
Host Species Mouse
Immunogen A recombinant protein of FSH-b subunit
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2488
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.