Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FSH beta Antibody (SPM107), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2451330.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
FSH beta Monoclonal antibody specifically detects FSH beta in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
FSH beta | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
P01225 | |
FSHB | |
Full length purified Human FSH | |
Protein A or G purified | |
RUO | |
Primary | |
This MAb reacts with a protein of 22kDa, identified as beta sub-unit of FSH. It does not cross react with the alpha sub-unit. Follicle stimulating hormone (FSH) is a hormone synthesized and secreted by gonadotrophs in the anterior pituitary gland. In the ovary, FSH stimulates the growth of immature Graafian follicles to maturation. As the follicle grows, it releases inhibin, which deactivates the FSH production. In men, FSH enhances the production of androgen-binding protein by the Sertoli cells of the testis and is critical for spermatogenesis. FSH and LH act synergistically in reproduction. FSH is a useful marker in the classification of pituitary tumors and the study of pituitary disease. | |
Store at 4C. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
SPM107 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
follicle stimulating hormone, beta polypeptide, Follicle-stimulating hormone beta subunit, Follitropin beta chain, follitropin subunit beta, follitropin, beta chain, FSH-B, FSH-beta | |
Mouse | |
22 kDa | |
0.2 mg | |
Cancer | |
2488 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction