Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FSH beta Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111651
Description
Immunogen sequence: TRDLVYKDPA RPKIQKTCTF KELVYETVRV PGCAHHADSL YTYPVATQCH CGK.
Human follicle stimulating hormone is a glycoprotein produced from urine of post-menopausal women and having a total molecular mass of 35,500 Dalton. HuFSH is a heterodimeric hormone consisting of 165 amino acids a chain (non-active) and 115 amino acids.
Specifications
FSH beta | |
Polyclonal | |
Unconjugated | |
FSHB | |
follicle stimulating hormone beta subunit; follicle stimulating hormone, beta polypeptide; follicle-stimulating hormone beta subunit; Follitropin beta chain; follitropin subunit beta; follitropin, beta chain; FSHB; FSH-B; FSH-beta; HH24 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2488 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P01225 | |
FSHB | |
Recombinant Protein corresonding to Human FSH-beta. Recombinant protein control fragment (Product #RP-107705). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction