Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSHB Mouse anti-Human, Mouse, Clone: P4G2, Abnova™

Catalog No. 89035464 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-464 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-464 Supplier Abnova Corporation Supplier No. MAB14059

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human FSHB.

  • High specificity
  • High reactivity

Specifications

Antigen FSHB
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone P4G2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human FSHB.
Dilution ELISA Immunocytochemistry (2 ug/mL) Immunofluorescence (2 ug/mL) Immunoprecipitation Radioimmunoassay Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 0.01 M potassium phosphate buffer, 2.5 mM KCl, 0.135 M NaCl (1 mg/mL BSA, 0.05% sodium azide).
Gene Accession No. P01225
Gene Symbols FSHB
Host Species Mouse
Immunogen Human FSHB.
Purification Method Ion exchange chromatography purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2488
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.