Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FSHR Antibody (626717), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB65591K
This item is not returnable.
View return policy
Description
FSHR Monoclonal antibody specifically detects FSHR in Human samples. It is validated for Flow CytometrySpecifications
FSHR | |
Monoclonal | |
DyLight 488 | |
follicle stimulating hormone receptor, follicle-stimulating hormone receptor, Follitropin receptor, FSH receptor, FSH-R, LGR1FSHRO, MGC141667, MGC141668, ODG1 | |
NS0 mouse myeloma cell line transfected with human FSH R, Accession # M65085 | |
0.5 mL | |
GPCR | |
2492 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry | |
626717 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction