Learn More
FSHR Antibody (6E8.2F5), HRP, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP236489H
Catalog No. NBP236489H
Only null left
Add to Cart
Description
Description
FSHR Monoclonal specifically detects FSHR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
FSHR
Monoclonal
HRP
PBS with No Preservative
follicle stimulating hormone receptor, follicle-stimulating hormone receptor, Follitropin receptor, FSH receptor, FSH-R, LGR1FSHRO, MGC141667, MGC141668, ODG1
Mouse
Protein G purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
6E8.2F5
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
P23945
FSHR
A synthetic peptide made to the human FSH R protein sequence (between residues 1-200). [Swiss-Prot P23945]
0.1 mL
GPCR
2492
Store at 4C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.