Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FSHR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA591952
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA591952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA591952 Supplier Invitrogen™ Supplier No. PA591952
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SW620; Cellular Location: Cell membrane, Multi-pass membrane protein Immunogen sequence: KNFRRDFFIL LSKCGCYEMQ AQIYRTETSS TVHNTHPRNG HCSSAPRVTS GSTYILVPLS HLAQN.

Follicle-Stimulating Hormone Receptor (FSHR) expression has been reported in ovary, uterus, and testis. ESTs have been isolated from testis libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSHR
Applications ELISA, Western Blot, Western Blot
Classification Polyclonal
Concentration 2.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FSHR
Gene Accession No. P20395, P23945, P35378
Gene Alias follicle stimulating hormone receptor; follicle-stimulating hormone receptor; Follitropin receptor; FSH receptor; Fshr; FSH-R; FSHR1; FSHRO; LGR1; ODG1
Gene Symbols FSHR
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 631-695 of human FSHR (NP_0001362).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14309, 2492, 25449
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.