Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FSIP1 Antibody (OTI4F9), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272451G
Description
FSIP1 Monoclonal specifically detects FSIP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FSIP1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FSIP1 | |
Full length recombinant protein of human FSIP1 (NP_689810) produced in HEK293T cells. | |
0.1 mL | |
161835.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4F9 | |
Western Blot, Immunohistochemistry | |
fibrous sheath interacting protein 1, fibrous sheath-interacting protein 1, FLJ35989 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction