Learn More
FSIP1 Mouse anti-Human, Clone: OTI4F9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806085

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The exact functions of FSIP1 remain unknown.Specifications
FSIP1 | |
Monoclonal | |
Unconjugated | |
FSIP1 | |
HSD10 | |
Mouse | |
Affinity Chromatography | |
RUO | |
161835 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4F9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8NA03 | |
FSIP1 | |
Full length human recombit protein of human FSIP1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.