Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FSIP1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30459820UL
Description
FSIP1 Polyclonal specifically detects FSIP1 in Mouse samples. It is validated for Western Blot.Specifications
FSIP1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
161835 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
fibrous sheath interacting protein 1, fibrous sheath-interacting protein 1, FLJ35989 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human FSIP1 (NP_689810.3). | |
20 μL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction