Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FSIP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112962
Description
FSIP2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
FSIP2 is a protein coding gene.
Specifications
| FSIP2 | |
| Polyclonal | |
| Unconjugated | |
| FSIP2 | |
| fibrous sheath interacting protein 2; fibrous sheath-interacting protein 2; FSIP2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 401024 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q5CZC0 | |
| FSIP2 | |
| Recombinant Human Fibrous sheath-interacting protein 2 protein (5530-5824AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction