Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FSTL3 Polyclonal Antibody

Catalog No. PIPA5121980
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121980 20 μL
PIPA5119126 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121980 Supplier Invitrogen™ Supplier No. PA5121980
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FSTL3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Follistatin-like 3 (FSTL3) is a gene located on chromosome 19p13.3 and encodes a secreted glycoprotein that belongs to the follistatin-related protein family. FSTL3 functions as an antagonist to members of the transforming growth factor-beta (TGF-β) superfamily, particularly inhibiting activin and myostatin signaling. This inhibitory action is crucial for regulating muscle growth, fertility, and inflammation. FSTL3 is expressed in various tissues, including the placenta, gonads, and skeletal muscle, where it modulates physiological processes by binding to and neutralizing the activities of its target ligands. Dysregulation of FSTL3 has been implicated in conditions such as muscle wasting, metabolic syndrome, and reproductive disorders, reflecting its role in maintaining tissue homeostasis and function. Research into FSTL3 focuses on its potential therapeutic applications in diseases involving TGF-β pathway dysregulation, offering insights into strategies for modulating growth factor activities in clinical settings.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSTL3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene FSTL3
Gene Accession No. Q9EQC7
Gene Alias E030038F23Rik; FLRG; FLRGFSRP; follistatin like 3; follistatin-like 3; follistatin-like 3 (secreted glycoprotein); follistatin-like protein 3; Follistatin-related gene protein; follistatin-related protein 3; FSRP; FSTL3; UNQ674/PRO1308
Gene Symbols FSTL3
Host Species Rabbit
Immunogen Recombinant protein Follistatin Like Protein 3. The antigen corresponds to amino acid range 24-256 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83554
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.