Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FSTL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579285
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579285 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579285 Supplier Invitrogen™ Supplier No. PA579285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human U2OS whole cell, human Hela whole cell, human A549 whole cell, human PC-3 whole cell, human HepG2 whole cell, human K562 whole cell, rat liver tissue, rat testis tissue, rat NRK whole cell, mouse liver tissue, mouse testis tissue, mouse lung tissue.

Follistatin-like 3 (FSTL3) is a gene located on chromosome 19p13.3 and encodes a secreted glycoprotein that belongs to the follistatin-related protein family. FSTL3 functions as an antagonist to members of the transforming growth factor-beta (TGF-β) superfamily, particularly inhibiting activin and myostatin signaling. This inhibitory action is crucial for regulating muscle growth, fertility, and inflammation. FSTL3 is expressed in various tissues, including the placenta, gonads, and skeletal muscle, where it modulates physiological processes by binding to and neutralizing the activities of its target ligands. Dysregulation of FSTL3 has been implicated in conditions such as muscle wasting, metabolic syndrome, and reproductive disorders, reflecting its role in maintaining tissue homeostasis and function. Research into FSTL3 focuses on its potential therapeutic applications in diseases involving TGF-β pathway dysregulation, offering insights into strategies for modulating growth factor activities in clinical settings.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSTL3
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene FSTL3
Gene Accession No. O95633, Q99PW7, Q9EQC7
Gene Alias E030038F23Rik; FLRG; FLRGFSRP; follistatin like 3; follistatin-like 3; follistatin-like 3 (secreted glycoprotein); follistatin-like protein 3; Follistatin-related gene protein; follistatin-related protein 3; FSRP; FSTL3; UNQ674/PRO1308
Gene Symbols FSTL3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human FSTL3 (145-159aa ECELRAARCRGHPDL).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10272, 114031, 83554
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.