Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FSTL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109739 Supplier Invitrogen™ Supplier No. PA5109739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GWMDPGTSRG PDVGVGESQA EEPRSFEVTR REGLSSHNEL LASCGKKFCS RGSRCVLSRK TGEPECQCLE ACRPSYVPVC GSDGRFYENH CKLHRAACLL GKRITVIHSK DCFLKGDTCT MAGYARLKNV LLALQTRLQP LQEGDSRQDP ASQKRLLVES LFRDLDADGN GHLSSSELAQ HVLKKQDLDE DLLGCSPGDL LRFDDYNSDS SLTLREFYMA FQVVQLSLA.

FSTL4 (Follistatin Like 4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includecalcium ion binding. An important paralog of this gene isFSTL5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSTL4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FSTL4
Gene Accession No. Q5STE3, Q6MZW2
Gene Alias B230374F23Rik; follistatin like 4; follistatin-like 4; follistatin-like protein 4; Follistatin-related protein 4; FSL4; FSTL4; KIAA1061; m-D/Bsp120I 1-1; mKIAA1061; SPIG1
Gene Symbols FSTL4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-250 of human FSTL4 (NP_0558971).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23105, 303130, 320027
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.