Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FSTL5 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300121R
Description
FSTL5 Polyclonal specifically detects FSTL5 in Human samples. It is validated for ELISA.Specifications
FSTL5 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
56884 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 550 | |
follistatin-like 5, Follistatin-like protein 5, follistatin-related protein 5, KIAA1263DKFZp566D234 | |
Produced in rabbits immunized with purified, recombinant FSTL5 (Q8N475; Gln32-Ala847). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction