Learn More
Invitrogen™ FSTL5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595795
Description
Immunogen sequence: RPTKEGGYGL KSYQPLMRLR HKQEKNQESS RVKGFMIQDG PFGSCENKYC GLGRHCVTSR ETGQAECACM DLCKRHYKPV CGSDGEFYEN HCEVHRAACL KKQKITIVHN EDCFFKGDKC KTTEYSKMKN MLLDLQNQKY IMQENENPNG DDISRKKLLV DQMFKYFDAD SNGLVDINEL TQVIKQEELG KDLFDCTLYV LLKYDDFNAD KHLALEEFYR AFQVIQLSLP; Positive Samples: SH-SY5Y, Mouse lung, Mouse brain, Rat testis.

Specifications
FSTL5 | |
Polyclonal | |
Unconjugated | |
FSTL5 | |
9130207J01Rik; follistatin like 5; follistatin-like 5; follistatin-like protein 5; Follistatin-related protein 5; Fstl5; Kiaa1263; m-D/Bsp120I 1-2; ollistatin-like 5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
213262, 365823, 56884 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.1 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8BFR2, Q8N475 | |
FSTL5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 21-250 of human FSTL5 (NP_0645012). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.