Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FTCD Monoclonal Antibody (OTI4C3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504945S


Description
FTCD Monoclonal antibody specifically detects FTCD in Human samples. It is validated for Western BlotSpecifications
FTCD | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O95954 | |
Mouse | |
Affinity chromatography | |
RUO | |
10841 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI4C3 | |
Unconjugated | |
Ftcd | |
58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1 | |
Full length human recombinant protein of human FTCD produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction