Learn More
FTCD Mouse anti-Human, Clone: OTI8C2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504981

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.Specifications
FTCD | |
Monoclonal | |
Unconjugated | |
FTCD | |
LCHC1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10841 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI8C2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O95954 | |
FTCD | |
Full length human recombit protein of human FTCD produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.