Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FTCDNL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145197 Supplier Invitrogen™ Supplier No. PA5145197
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HVPGCSVFLF GEADLPEKRS LVQRRKQLGW FTRRDFSALQ PDLGAAPSQR CGLTACFRAL Antigen sequence identity: Mouse 26%; Rat 28%.

Formiminotransferase Cyclodeaminase N-Terminal Like (FTCDNL1) is a protein coding gene predicted to enable folic acid binding activity and transferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FTCDNL1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FTCDNL1
Gene Accession No. E5RQL4
Gene Alias FONG; Formiminotransferase Cyclodeaminase N-Terminal Like
Gene Symbols FTCDNL1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 78-117. Recombinant protein control fragment (Product #RP-109858).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348751
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.