Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FTHL17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569658
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569658 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569658 Supplier Invitrogen™ Supplier No. PA569658
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

This gene encodes a ferritin heavy chain-like protein. This gene is primarily expressed in embryonic germ cells. The encoded protein may lack ferroxidase activity. Multiple pseudogenes of this gene are found on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FTHL17
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FTHL17
Gene Accession No. Q9BXU8
Gene Alias cancer/testis antigen 38; CT38; ferritin heavy chain like 17; ferritin heavy polypeptide-like 17; ferritin, heavy polypeptide-like 17; FTHL17
Gene Symbols FTHL17
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human FTHL17.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53940
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.