Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTL Mouse anti-Human, Unconjugated, Clone: FTL/1387, Abnova™

Catalog No. 89337488 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-488 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-488 Supplier Abnova Corporation Supplier No. MAB20995
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

FTL Monoclonal antibody specifically detects FTL in Human samples. It is validated for WB, IHC-P.

Specifications

Antigen FTL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FTL/1387
Conjugate Unconjugated
Description ferritin, light polypeptide
Gene FTL
Gene Alias MGC71996
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 38-165 of human FTL
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2512
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.