Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FTMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530906
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530906 Supplier Invitrogen™ Supplier No. PA530906
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (87%), Rat (87%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

FTMT stores iron in a soluble, non-toxic, readily available form. FTMT is important for iron homeostasis and has ferroxidase activity. Iron is taken up in the ferrous form and deposited as ferric hydroxides after oxidation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FTMT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7
Gene Ftmt
Gene Accession No. Q8N4E7, Q9D5H4
Gene Alias 4930447C24Rik; FerH; ferritin H; ferritin H subunit; ferritin heavy chain 3; ferritin mitochondrial; ferritin, mitochondrial; Fth3; FTMT; mitochondrial ferritin; MtF
Gene Symbols Ftmt
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Mitochondrial Ferritin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67634, 94033
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.