Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FTO Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11060935AF350
Description
FTO Polyclonal specifically detects FTO in Human, Mouse, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoblotting.Specifications
FTO | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunoblotting | |
alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
Rabbit | |
Affinity Purified | |
Lipid and Metabolism, Neuroscience | |
79068.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunoblot | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
FTO | |
A synthetic peptide within the C-terminal region (residues 400-505) of the human FTO protein. [Swiss-ProtNo. Q9C0B1] | |
0.1 mL | |
Primary | |
Human, Mouse, Chicken, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction