Learn More
FTO Mouse anti-Human, Clone: OTI4A1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809392

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Rising obesity rates are rapidly becoming a growing health concern in the developing world. The fat mass and obesity associated gene (FTO) is the first gene discovered to contribute to common forms of human obesity. FTO is a member of the non-heme dioxygenase superfamily, encoding a 2-oxoglutarate-dependent nucleic acid demethylase whose mRNA is widely expressed, especially in neurons of feeding-related nuclei of the brain. FTO mRNA in the arcuate nucleus in mice is up-regulated by feeding and down-regulated during fasting, although the opposite pattern has been observed in rats. At least four isoforms of FTO are known to exist.Specifications
FTO | |
Monoclonal | |
Unconjugated | |
FTO | |
ALKBH9, BMIQ14, GDFD | |
Mouse | |
Affinity Chromatography | |
RUO | |
79068 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4A1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9C0B1 | |
FTO | |
Full length human recombit protein of human FTO produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.