Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FTO, Mouse anti-Human,Mouse,Rat, Clone: FT86-4, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280045100UG
Description
FTO Monoclonal specifically detects FTO in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation.Specifications
FTO | |
Monoclonal | |
1 mg/mL | |
Western Blot, ELISA, Immunoprecipitation | |
alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Recognizes human, mouse and rat FTO. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, ELISA, Immunoprecipitation | |
FT86-4 | |
Unconjugated | |
0.2 um-filtered solution in PBS, pH 7.4 | |
FTO | |
Recombinant human FTO. | |
100 μg | |
Lipid and Metabolism, Neuroscience | |
79068 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction