Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTO, Mouse anti-Mouse, Clone: FT62-6, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP28005050UG

 View more versions of this product

Catalog No. NB013177


Only null left
Add to Cart

Description

Description

FTO Monoclonal specifically detects FTO in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

FTO
Monoclonal
1 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149
Mouse
Protein G purified
RUO
Primary
Recognizes mouse FTO. Weakly cross-reacts with human FTO. Does not cross-react with rat FTO.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
FT62-6
Unconjugated
0.2 um-filtered solution in PBS, pH 7.4
FTO
Recombinant human FTO.
50 μg
Lipid and Metabolism, Neuroscience
79068
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.