Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FTO Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA146310
Description
Suggested positive control: human brain lysate, human brain protein.
FTO is a novel protein and a member of the non-heme dioxygenase (Fe(II)-and 2-oxoglutarate-dependent dioxygenases) superfamily. Amino acid conservation patterns support this hypothesis and indicates that both 2-oxoglutarate and iron should be important for FTO function. Though not much is yet known about FTO, it is thought that the protein may somehow modify the activity of genes involved in metabolism and fat storage, which in turn may influence a person's risk of obesity.
Specifications
FTO | |
Polyclonal | |
Unconjugated | |
Fto | |
AlkB homolog 9; ALKBH9; alpha-ketoglutarate-dependent dioxygenase FTO; AW743446; BMIQ14; fat mass and obesity associated; fat mass and obesity-associated protein; fatso; FTO; GDFD; KIAA1752; m6A(m)-demethylase FTO; MGC5149; mKIAA1752; mRNA (2'-O-methyladenosine-N(6)-)-demethylase FTO; mRNA N(6)-methyladenosine demethylase FTO; Protein fatso; protein fto; RGD1305121; tRNA N1-methyl adenine demethylase FTO; U6 small nuclear RNA (2'-O-methyladenosine-N(6)-)-demethylase FTO; U6 small nuclear RNA N(6)-methyladenosine-demethylase FTO | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26383, 415718, 79068 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
1.1 mg/mL | |
Tris citrate, Tris phosphate with 0.1% sodium azide; pH 7-8 | |
Q8BGW1, Q9C0B1 | |
Fto | |
Synthetic peptide corresponding to an C-terminal portion within residues 400-505 of the human protein. [Swiss-Prot# Q9C0B1]. | |
100 μL | |
Primary | |
Human, Mouse, Monkey, Chicken | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction