Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FTO Rabbit anti-Human, Clone: S04-1F7, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | FTO |
---|---|
Clone | S04-1F7 |
Dilution | Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
FTO Monoclonal antibody specifically detects FTO in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
FTO | |
Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human | |
alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
A synthetic peptide of human FTO (Uniprot#: Q9C0B1) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S04-1F7 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Neuroscience | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
79068 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title