Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FTO Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | FTO |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5611
![]() ![]() |
Proteintech Group Inc
272261AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Rising obesity rates are rapidly becoming a growing health concern in the developing world. The fat mass and obesity associated gene (FTO) is the first gene discovered to contribute to common forms of human obesity. FTO is a member of the non-heme dioxygenase superfamily, encoding a 2-oxoglutarate-dependent nucleic acid demethylase whose mRNA is widely expressed, especially in neurons of feeding-related nuclei of the brain. FTO mRNA in the arcuate nucleus in mice is up-regulated by feeding and down-regulated during fasting, although the opposite pattern has been observed in rats. At least four isoforms of FTO are known to exist.Specifications
FTO | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8BGW1, Q9C0B1 | |
26383, 79068 | |
FTO Fusion Protein Ag26095 | |
Primary | |
-20°C | |
FTO |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FTO, KIAA1752 | |
FTO | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title