Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FTO Recombinant Rabbit Monoclonal Antibody (4A5T6)
GREENER_CHOICE

Catalog No. FEMA551209
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551209 Supplier Invitrogen™ Supplier No. MA551209
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: STGTLDYILQ RCQLALQNVC DDVDNDDVSL KSFEPAVLKQ GEEIHNEVEF EWLRQFWFQG NRYRKCTDWW CQPMAQLEAL WKKMEGVTNA VLHEVKREGL PVEQRNEILT AILASLTARQ NLRREWHARC QSRIARTLPA DQKPECRPYW EKDDASMPLP FDLTDIVSEL RGQLLEAK.

FTO is a novel protein and a member of the non-heme dioxygenase (Fe(II)-and 2-oxoglutarate-dependent dioxygenases) superfamily. Amino acid conservation patterns support this hypothesis and indicates that both 2-oxoglutarate and iron should be important for FTO function. Though not much is yet known about FTO, it is thought that the protein may somehow modify the activity of genes involved in metabolism and fat storage, which in turn may influence a person's risk of obesity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FTO
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 4A5T6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Fto
Gene Accession No. Q2A121, Q8BGW1, Q9C0B1
Gene Alias AlkB homolog 9; ALKBH9; alpha-ketoglutarate-dependent dioxygenase FTO; AW743446; BMIQ14; fat mass and obesity associated; fat mass and obesity-associated protein; fatso; FTO; GDFD; KIAA1752; m6A(m)-demethylase FTO; MGC5149; mKIAA1752; mRNA (2'-O-methyladenosine-N(6)-)-demethylase FTO; mRNA N(6)-methyladenosine demethylase FTO; Protein fatso; protein fto; RGD1305121; tRNA N1-methyl adenine demethylase FTO; U6 small nuclear RNA (2'-O-methyladenosine-N(6)-)-demethylase FTO; U6 small nuclear RNA N(6)-methyladenosine-demethylase FTO
Gene Symbols Fto
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 327-504 of human FTO (NP_001073901.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26383, 291905, 79068
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.