Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTSJ1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB293163 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293163 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293163 Supplier Novus Biologicals Supplier No. NBP326088C
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FTSJ1 Polyclonal antibody specifically detects FTSJ1 in Human samples. It is validated for Western Blot, Immunocytochemistry

Specifications

Antigen FTSJ1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias CDLIV, cell division protein, EC 2.1.1.-, FtsJ homolog 1 (E. coli), JM23, mental retardation, X-linked 44, mental retardation, X-linked 9, MRX44, Protein ftsJ homolog 1, putative ribosomal RNA methyltransferase 1, rRNA (uridine-2'-O-)-methyltransferase, SPB1MRX9, TRM7
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FTSJ1
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24140
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.