Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FTSJ1 Polyclonal Antibody

Catalog No. PIPA535886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535886 Supplier Invitrogen™ Supplier No. PA535886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG, SK-N-SH, IMR32, SK-N-AS. Predicted reactivity: Mouse (87%), Pig (90%), Rhesus Monkey (97%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the S-adenosylmethionine-binding protein family. It is a nucleolar protein and may be involved in the processing and modification of rRNA. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FTSJ1
Applications Western Blot
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene FTSJ1
Gene Accession No. Q9UET6
Gene Alias 2'-O-ribose RNA methyltransferase TRM7 homolog; AI931847; CDLIV; cell division protein; fb17g12; Ftsj; Ftsj homolog 1; FtsJ homolog 1 (E. coli); FtsJ RNA 2'-O-methyltransferase 1; FtsJ RNA methyltransferase homolog 1; FtsJ RNA methyltransferase homolog 1 (E. coli); FTSJ1; Ftsjl; FtsJ-like protein 1; JM23; MRX44; MRX9; Protein ftsJ homolog 1; putative ribosomal RNA methyltransferase 1; putative tRNA (cytidine(32)/guanosine(34)-2'-O)-methyltransferase; RGD1561061; rRNA (uridine-2'-O-)-methyltransferase; Sfc12; SPB1; TRM7; TRMT7; tRNA methyltransferase 7 homolog; wu:fb17g12; zgc:101831
Gene Symbols FTSJ1
Host Species Rabbit
Immunogen Full length human FTSJ1 Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24140
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.