Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTSJ1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982087
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2087 Supplier Bioss Supplier No. BS12264R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of the S-adenosylmethionine-binding protein family. It is a nucleolar protein and may be involved in the processing and modification of rRNA. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.

Specifications

Antigen FTSJ1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FTSJ1
Gene Accession No. Q9UET6
Gene Alias 2′-O-ribose RNA methyltransferase TRM7 homolog; AI931847; BOS_25929; CDLIV; cell division protein; fb17g12; Ftsj; Ftsj homolog 1; FtsJ homolog 1 (E. coli); FtsJ RNA methyltransferase homolog 1; FtsJ RNA methyltransferase homolog 1 (E. coli); FTSJ1; Ftsjl; FtsJ-like protein 1; JM23; LOC100353966; MRX44; MRX9; Protein ftsJ homolog 1; putative ribosomal RNA methyltransferase 1; Putative tRNA (cytidine(32)/guanosine(34)-2′-O)-methyltransferase; RGD1561061; rRNA (uridine-2′-O-)-methyltransferase; Sfc12; SPB1; TRM7; TRMT7; tRNA methyltransferase 7 homolog; wu:fb17g12; zgc:101831
Gene Symbols FTSJ1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FTSJ1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24140
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.