Learn More
Invitrogen™ FTSJ2 Polyclonal Antibody

Description
Immunogen sequence: DPSSPVGFVL GVDLLHIFPL EGATFLCPAD VTDPRTSQRI LEVLPGRRAD VILSDMAPNA TGFRDLDHDR LI Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Specifications
Specifications
| Antigen | FTSJ2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MRM2 |
| Gene Accession No. | Q9UI43 |
| Gene Alias | 16S rRNA (uridine(1369)-2'-O)-methyltransferase; 16S rRNA [Um1369] 2'-O-methyltransferase; 2310037B18Rik; cell division protein FtsJ; epididymis luminal protein 97; FJH1; FtsJ homolog 2; FtsJ homolog 2 (E. coli); FtsJ RNA methyltransferase homolog 2; FTSJ2; HEL97; mitochondrial rRNA methyltransferase 2; Mrm2; MRM2 RNA methyltransferase homolog; protein ftsJ homolog 2; putative ribosomal RNA methyltransferase 2; RRMJ2; rRNA (uridine-2'-O-)-methyltransferase; rRNA methyltransferase 2, mitochondrial |
| Gene Symbols | MRM2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.