Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FTSJ2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562021 Supplier Invitrogen™ Supplier No. PA562021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LCLTLLSVTP DILQPGGTFL CKTWAGSQSR RLQRRLTEEF QNVRIIKPEA SRKESSEVYF LATQYHGRKG TVKQ Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 70%.

Putative ribosomal RNA methyltransferase 2 (FTSJ2), also named as FJH1 or Protein FTSJ homolog 2 is a 246 amino acid protein, which belongs to the methyl-transferase superfamily. FTSJ2 localizes in the nucleus and is widely expressed in in muscle, placenta, and heart. FTSJ2 functions as a nucleolar RNA methyl-transferase involved in eukaryotic RNA processing and modification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FTSJ2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRM2
Gene Accession No. Q9UI43
Gene Alias 16S rRNA (uridine(1369)-2'-O)-methyltransferase; 16S rRNA [Um1369] 2'-O-methyltransferase; 2310037B18Rik; cell division protein FtsJ; epididymis luminal protein 97; FJH1; FtsJ homolog 2; FtsJ homolog 2 (E. coli); FtsJ RNA methyltransferase homolog 2; FTSJ2; HEL97; mitochondrial rRNA methyltransferase 2; Mrm2; MRM2 RNA methyltransferase homolog; protein ftsJ homolog 2; putative ribosomal RNA methyltransferase 2; RRMJ2; rRNA (uridine-2'-O-)-methyltransferase; rRNA methyltransferase 2, mitochondrial
Gene Symbols MRM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FTSJ2. Recombinant protein control fragment (Product #RP-92697).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29960
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.