Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FTSJ3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569555
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569555 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569555 Supplier Invitrogen™ Supplier No. PA569555
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

Although the function of this gene is not known, the existence of this gene is supported by mRNA and EST data. A possible function of the encoded protein can be inferred from amino acid sequence similarity to the E.coli FtsJ protein and to a mouse protein possibly involved in embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FTSJ3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FTSJ3
Gene Accession No. Q5RJT2
Gene Alias 2'-O-ribose RNA methyltransferase SPB1 homolog; 2'-O-ribose RNA methyltransferase SPB1 homolog {ECO:0000255; AA537063; AU045295; C79843; D11Ertd400e; ectoplacental cone, invasive trophoblast giant cells, extraembryonic ectoderm and chorion sequence 3; Epcs3; FLJ20062; FtsJ homolog 3; FtsJ homolog 3 (E. coli); FtsJ RNA 2'-O-methyltransferase 3; FtsJ RNA methyltransferase homolog 3; FTSJ3; HAMAP-Rule:MF_03163}; pre-rRNA 2'-O-ribose RNA methyltransferase FTSJ3; pre-rRNA 2'-O-ribose RNA methyltransferase FTSJ3; pre-rRNA processing protein FTSJ3; pre-rRNA processing protein FTSJ3; pre-rRNA processing protein FTSJ3 {ECO:0000255; protein ftsJ homolog 3; Putative rRNA methyltransferase 3; putative rRNA methyltransferase 3 {ECO:0000255; rRNA (uridine-2'-O-)-methyltransferase 3; SB92; SPB1; SPB1 RNA methyltransferase homolog
Gene Symbols FTSJ3
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of rat FTSJ3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303608
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.