Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FTSJD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561696
Description
Immunogen sequence: DKVAKGYFNS WAEEHGVYHP GQSSILEGTA SNLECHLWHI LEGKKLPKVK CSPFCNGEIL KTLNEAIEKS LGGAFNLDSK FRPKQQYSCS CH Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 75%.
S-adenosyl-L-methionine-dependent methyltransferase that mediates mRNA cap2 2'-O-ribose methylation to the 5'-cap structure of mRNAs. Methylates the ribose of the second nucleotide of a m(7)GpppG-capped mRNA and small nuclear RNA (snRNA) (cap0) to produce m(7)GpppRmpNm (cap2). Recognizes a guanosine cap on RNA independently of its N(7) methylation status. Display cap2 methylation on both cap0 and cap1. Displays a preference for cap1 RNAs.
Specifications
FTSJD1 | |
Polyclonal | |
Unconjugated | |
CMTR2 | |
adrift homolog; AFT; AU022703; C730036L12Rik; cap methyltransferase 2; cap2 2'O-ribose methyltransferase 2; Cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 2; Cmtr2; FtsJ methyltransferase domain containing 1; ftsJ methyltransferase domain-containing protein 1; ftsjd1; HMTr2; hypothetical protein LOC406742; MTr2; Protein adrift homolog; RGD1309394; wu:fj48c05; zgc:55686 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55783 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IYT2 | |
CMTR2 | |
Recombinant protein corresponding to Human FTSJD1. Recombinant protein control fragment (Product #RP-98778). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction