Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTSJD2 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB248493 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB248493 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB248493 Supplier Novus Biologicals Supplier No. NB10079786B
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FTSJD2 Polyclonal antibody specifically detects FTSJD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen FTSJD2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Cap1 2'O-ribose methyltransferase 1, cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1, EC 2.1.1.57, FtsJ methyltransferase domain containing 2, FtsJ methyltransferase domain-containing protein 2, hMTr1, Interferon-stimulated gene 95 kDa protein, ISG95, KIAA0082FLJ22156, MTR1, S-adenosyl-L-methionine-dependent methyltransferase FTSJD2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human KIAA0082 using the numbering given in TrEMBL entry Q8N1G2 (GeneID 23070).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23070
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.