Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FTSJD2 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079786H
Description
FTSJD2 Polyclonal antibody specifically detects FTSJD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
FTSJD2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Cap1 2'O-ribose methyltransferase 1, cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1, EC 2.1.1.57, FtsJ methyltransferase domain containing 2, FtsJ methyltransferase domain-containing protein 2, hMTr1, Interferon-stimulated gene 95 kDa protein, ISG95, KIAA0082FLJ22156, MTR1, S-adenosyl-L-methionine-dependent methyltransferase FTSJD2 | |
The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human KIAA0082 using the numbering given in TrEMBL entry Q8N1G2 (GeneID 23070). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23070 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction