Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FTSJD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556316
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556316 Supplier Invitrogen™ Supplier No. PA556316
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQGRKDIVEA SSQKGRRGLG LTLRGFDQEL NVDWRDEPEP SACEQVSWFP ECTTEIPDTQ EMSDWMVVGK RKMIIEDETE FCGEELLHSV LQCK Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

S-adenosyl-L-methionine-dependent methyltransferase that mediates mRNA cap1 2'-O-ribose methylation to the 5'-cap structure of mRNAs. Methylates the ribose of the first nucleotide of a m(7)GpppG-capped mRNA and small nuclear RNA (snRNA) to produce m(7)GpppRm (cap1). Displays a preference for cap0 transcripts. Cap1 modification is linked to higher levels of translation. May be involved in the interferon response pathway. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FTSJD2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CMTR1
Gene Accession No. Q5U2Z5, Q8N1G2, Q9DBC3
Gene Alias 1300018I05Rik; AI451264; Cap methyltransferase 1; Cap1 2'O-ribose methyltransferase 1; cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1; cmtr1; FtsJ methyltransferase domain containing 2; ftsJ methyltransferase domain-containing protein 2; ftsjd2; hMTr1; hypothetical protein LOC393102; interferon-stimulated gene 95 kDa protein; ISG95; KIAA0082; LOW QUALITY PROTEIN: cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1; LRRG00129; mKIAA0082; MTr1; RGD1307801; S-adenosyl-L-methionine-dependent methyltransferase ftsjd2; zgc:55336
Gene Symbols CMTR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FTSJD2. Recombinant protein control fragment (Product #RP-94860).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23070, 309656, 74157
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.