Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ FTY 720
GREENER_CHOICESDP

Catalog No. 617650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-765-0 50 mg
61-761-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-765-0 Supplier R&D Systems™ Supplier No. 6176/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent S1P receptor agonist; also immunosuppressant

FTY 720 is an S1P receptor agonist; undergoes phosphorylation by sphingosine kinase 2, which then binds S1P1,3,4 and 5 receptors (EC50 values are 0.3 - 0.6 nM for S1P1, 4 and 5 and ∼3 nM for S1P3). Immunosuppressant; sequesters lymphocytes in lymph nodes, reducing autoimmune response. Suppresses the development of EAE (an in vivo model of MS). Orally bioavailable and blood-brain barrier permeable.

Specifications

CAS 162359-56-0
Quantity 50 mg
Product Line Tocris
Target Sphingosine-1-phosphate Receptor
Molecular Formula C19H33NO2.HCl
Formula Weight 343.93g/mol
Product Type Agonist

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.