Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUBP1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP226146
Description
FUBP1 Polyclonal specifically detects FUBP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Peptide ELISA.Specifications
FUBP1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Fubp1 | |
Peptide with sequence C-DQIVEKGRPAPGFHH corresponding to internal region according to NP_476513.2. | |
0.1 mg | |
DNA replication Transcription Translation and Splicing | |
8880 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, ELISA | |
0.5 mg/mL | |
Western Blot 0.03 - 0.1 ug/ml, Immunohistochemistry 4 - 8 ug/ml, Peptide ELISA Detection limit 1:128000 | |
far upstream element (FUSE) binding protein 1, far upstream element binding protein, far upstream element-binding protein 1, FBPDNA helicase V, FUBP, FUSE-binding protein 1, hDH V | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction