Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUBP1 Mouse anti-Human, Clone: 14F5, Novus Biologicals™

Mouse Monoclonal Antibody
$340.50 - $499.50
Specifications
Antigen | FUBP1 |
---|---|
Clone | 14F5 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1053, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25944850
![]() |
Novus Biologicals
NBP25944850UL |
50 μL |
Each for $340.50
|
|
|||||
NBP259448X
![]() |
Novus Biologicals
NBP259448100UL |
100 μL |
Each for $499.50
|
|
|||||
Description
FUBP1 Monoclonal specifically detects FUBP1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
FUBP1 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
far upstream element (FUSE) binding protein 1, far upstream element binding protein, far upstream element-binding protein 1, FBPDNA helicase V, FUBP, FUSE-binding protein 1, hDH V | |
FUBP1 | |
IgG2b κ | |
Protein A purified |
14F5 | |
Western Blot 1:1000, Flow Cytometry 1:10-1:1053, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
8880 | |
Recombinant human FUBP1 (279-448aa) purified from E.coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title